Tisk…
3-(4-(Trifluoromethyl)phenyl)piperidin-3-ol hydrochloride
Dodavatel: Apollo Scientific
PC402144-1G PC402144-250MG
APOSPC402144-1GEA 18190 CZK
APOSPC402144-1G APOSPC402144-250MG
3-(4-(Trifluoromethyl)phenyl)piperidin-3-ol hydrochloride
3-(4-(Trifluoromethyl)phenyl)piperidin-3-ol hydrochloride
Vzorec: C₁₂H₁₄F₃NO·ClH
MW: 281.702 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28108755
CAS číslo: 2409589-75-7