6-Fluoro-7-nitro-3,4-dihydroisoquinolin-1(2H)-one
Dodavatel: Apollo Scientific
PC402140-1G
PC402140-250MG
APOSPC402140-1GEA
22750
CZK
APOSPC402140-1G
APOSPC402140-250MG
6-Fluoro-7-nitro-3,4-dihydroisoquinolin-1(2H)-one
6-Fluoro-7-nitro-3,4-dihydroisoquinolin-1(2H)-one
Vzorec:
C₉H₇FN₂O₃ MW: 210,162 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD31617681 CAS číslo: 1809018-61-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...