Tisk…

3-(Aminomethyl)-3-(2-fluorophenyl)cyclobutan-1-one hydrochloride

Dodavatel: Apollo Scientific

PC402136-1G PC402136-250MG
APOSPC402136-1GEA 22750 CZK
APOSPC402136-1G APOSPC402136-250MG
3-(Aminomethyl)-3-(2-fluorophenyl)cyclobutan-1-one hydrochloride
3-(Aminomethyl)-3-(2-fluorophenyl)cyclobutan-1-one hydrochloride
Vzorec: C₁₁H₁₂FNO·ClH
MW: 229,678 g/mol
Storage Temperature: Refrigerator
CAS číslo: 2409597-10-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR