4-Chloro-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidine
Dodavatel: Apollo Scientific
PC402135-1G
PC402135-250MG
PC402135-500MG
APOSPC402135-1GEA
22290
CZK
APOSPC402135-1G
APOSPC402135-250MG
APOSPC402135-500MG
4-Chloro-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidine
4-Chloro-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidine
Vzorec:
C₈H₄ClF₃N₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD28506297 CAS číslo: 1628317-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...