Tisk…

1-(Isoquinolin-5-yl)-3-(4-(trifluoromethyl)benzyl)urea 97%

Dodavatel: Apollo Scientific

PC402132-1G PC402132-500MG
APOSPC402132-1GEA 48930 CZK
APOSPC402132-1G APOSPC402132-500MG
1-(Isoquinolin-5-yl)-3-(4-(trifluoromethyl)benzyl)urea 97%
1-(Isoquinolin-5-yl)-3-(4-(trifluoromethyl)benzyl)urea
Vzorec: C₁₈H₁₄F₃N₃O
MW: 345,318 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07696058
CAS číslo: 581809-67-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR