4-(4-(Trifluoromethyl)phenyl)piperidine
Dodavatel: Apollo Scientific
PC402123-100MG
PC402123-1G
PC402123-250MG
APOSPC402123-100MGEA
8300
CZK
APOSPC402123-100MG
APOSPC402123-1G
APOSPC402123-250MG
4-(4-(Trifluoromethyl)phenyl)piperidine
4-(4-(Trifluoromethyl)phenyl)piperidine
Vzorec:
C₁₂H₁₄F₃N MW: 229,241 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05189944 CAS číslo: 67259-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...