Tisk…

2-Fluoro-1-isocyanato-4-methylbenzene 95%

Dodavatel: Apollo Scientific
Varování

PC402115-1G PC402115-250MG
APOSPC402115-1GEA 21300 CZK
APOSPC402115-1G APOSPC402115-250MG
2-Fluoro-1-isocyanato-4-methylbenzene 95%
2-Fluoro-1-isocyanato-4-methylbenzene

May form small amount of sediment

Vzorec: C₈H₆FNO
Storage Temperature: Refrigerator
MDL Number: MFCD18269662
CAS číslo: 69941-50-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR