Tisk…

2-(Trifluoromethyl)-1H-benzimidazole-7-carbaldehyde

Dodavatel: Apollo Scientific
Varování

PC402002-1G PC402002-250MG
APOSPC402002-1GEA 28570 CZK
APOSPC402002-1G APOSPC402002-250MG
2-(Trifluoromethyl)-1H-benzimidazole-7-carbaldehyde
2-(Trifluoromethyl)-1H-benzimidazole-7-carbaldehyde
Vzorec: C₉H₅F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD25460304
CAS číslo: 933741-41-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR