Tisk…

Ethyl 2-[3-chloro-2-oxo-5-(trifluoromethyl)pyridin-1-yl]acetate

Dodavatel: Apollo Scientific

PC401033-1G PC401033-5G
APOSPC401033-1GEA 8700 CZK
APOSPC401033-1G APOSPC401033-5G
Ethyl 2-[3-chloro-2-oxo-5-(trifluoromethyl)pyridin-1-yl]acetate
Ethyl 2-[3-chloro-2-oxo-5-(trifluoromethyl)pyridin-1-yl]acetate
Vzorec: C₁₀H₉ClF₃NO₃
MW: 283,632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28122854
CAS číslo: 1672655-84-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR