Tisk…

6-Chloro-2-(4-tert-butoxycarbonylpiperazino)-4-(trifluoromethyl)pyridine

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 4-(6-chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine-1-carboxylate, 6-Chloro-2-(4-tert-butoxycarbonylpiperazin-1-yl)-4-(trifluoromethyl)pyridine

PC400603-1G PC400603-5G
APOSPC400603-1GEA 4770 CZK
APOSPC400603-1G APOSPC400603-5G
6-Chloro-2-(4-tert-butoxycarbonylpiperazino)-4-(trifluoromethyl)pyridine
6-Chloro-2-(4-tert-butoxycarbonylpiperazino)-4-(trifluoromethyl)pyridine
Vzorec: C₁₅H₁₉ClF₃N₃O₂
MW: 365,778 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23159175
CAS číslo: 1201675-03-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR