Tisk…

Racemic cis-2-Fluorocyclopropanecarboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: cis-2-Fluorocyclopropanecarboxylic acid

PC400540-1G PC400540-5G
APOSPC400540-1GEA 6620 CZK
APOSPC400540-1G APOSPC400540-5G
Racemic cis-2-Fluorocyclopropanecarboxylic acid 95%
Racemic cis-2-Fluorocyclopropanecarboxylic acid
Vzorec: C₄H₅FO₂
MW: 104,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972869
CAS číslo: 105919-34-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR