Tisk…

(1S,2S)-2-Fluorocyclopropanecarboxylic acid 95+%

Dodavatel: Apollo Scientific
Varování

PC400537-1G PC400537-250MG
APOSPC400537-1GEA 1120 CZK
APOSPC400537-1G APOSPC400537-250MG
(1S,2S)-2-Fluorocyclopropanecarboxylic acid 95+%
(1S,2S)-2-Fluorocyclopropanecarboxylic acid
Vzorec: C₄H₅FO₂
MW: 104.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19237459
CAS číslo: 127199-14-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR