Tisk…

4-Amino-2-chloro-3-fluoropyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-3-fluoropyridin-4-amine

PC400526-1G PC400526-250MG
APOSPC400526-1GEA 2650 CZK
APOSPC400526-1G APOSPC400526-250MG
4-Amino-2-chloro-3-fluoropyridine
4-Amino-2-chloro-3-fluoropyridine
Vzorec: C₅H₄ClFN₂
MW: 146,55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606117
CAS číslo: 1227577-03-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR