Tisk…

1-(Azetidin-3-yl)-4-fluoropiperidine dihydrochloride

Dodavatel: Apollo Scientific

Synonyms: 3-(4-Fluoropiperidin-1-yl)azetane dihydrochloride, 3-(4-Fluoropiperidin-1-yl)azetidine dihydrochloride

PC400509-250MG PC400509-500MG
APOSPC400509-250MGEA 15740 CZK
APOSPC400509-250MG APOSPC400509-500MG
1-(Azetidin-3-yl)-4-fluoropiperidine dihydrochloride
1-(Azetidin-3-yl)-4-fluoropiperidine dihydrochloride
Vzorec: C₈H₁₅FN₂·2ClH
MW: 231,138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11867903
CAS číslo: 194427-15-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR