Tisk…

6-Bromo-2,3-difluoropyridine 95%

Dodavatel: Apollo Scientific
Varování

PC400179-1G PC400179-250MG
APOSPC400179-1GEA 34720 CZK
APOSPC400179-1G APOSPC400179-250MG
6-Bromo-2,3-difluoropyridine 95%
6-Bromo-2,3-difluoropyridine
Vzorec: C₅H₂BrF₂N
MW: 193,977 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20527926
CAS číslo: 1257071-45-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR