Tisk…

5-Bromo-3-fluoro-2-(methylsulphonyl)pyridine

Dodavatel: Apollo Scientific
Varování

PC400174-1G PC400174-250MG
APOSPC400174-1GEA 39680 CZK
APOSPC400174-1G APOSPC400174-250MG
5-Bromo-3-fluoro-2-(methylsulphonyl)pyridine
5-Bromo-3-fluoro-2-(methylsulphonyl)pyridine
Vzorec: C₆H₅BrFNO₂S
Storage Temperature: Ambient
MDL Number: MFCD28656592
CAS číslo: 1446002-35-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR