5-Bromo-6-fluoroisoquinoline
Dodavatel: Apollo Scientific
|
Varování
|
PC400167-1G
PC400167-250MG
APOSPC400167-1GEA
9360
CZK
APOSPC400167-1G
APOSPC400167-250MG
5-Bromo-6-fluoroisoquinoline
5-Bromo-6-fluoroisoquinoline
Vzorec:
C₉H₅BrFN MW: 226,045 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20527917 CAS číslo: 1239463-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...