5-Bromo-2-(difluoromethoxy)pyridine-3-carbaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-2-(difluoromethoxy)nicotinaldehyde
PC400164-100MG
PC400164-250MG
APOSPC400164-100MGEA
6090
CZK
APOSPC400164-100MG
APOSPC400164-250MG
5-Bromo-2-(difluoromethoxy)pyridine-3-carbaldehyde
5-Bromo-2-(difluoromethoxy)pyridine-3-carbaldehyde
Vzorec:
C₇H₄BrF₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD31621146 CAS číslo: 2231674-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...