Tisk…

(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC400162-100MG PC400162-250MG
APOSPC400162-100MGEA 5630 CZK
APOSPC400162-100MG APOSPC400162-250MG
(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanamine hydrochloride
(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanamine hydrochloride
Vzorec: C₇H₇BrF₂N₂O·ClH
MW: 289,505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621144
CAS číslo: 2231675-53-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR