5-Bromo-2-(difluoromethoxy)pyridine-3-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC400161-250MG
PC400161-100MG
APOSPC400161-250MGEA
11180
CZK
APOSPC400161-250MG
APOSPC400161-100MG
5-Bromo-2-(difluoromethoxy)pyridine-3-carbonitrile
5-Bromo-2-(difluoromethoxy)pyridine-3-carbonitrile
Vzorec:
C₇H₃BrF₂N₂O MW: 249,012 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28734738 CAS číslo: 1805580-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...