Tisk…

(2-(Difluoromethoxy)pyridin-3-yl)methanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC400158-100MG PC400158-250MG
APOSPC400158-100MGEA 6220 CZK
APOSPC400158-100MG APOSPC400158-250MG
(2-(Difluoromethoxy)pyridin-3-yl)methanamine hydrochloride
(2-(Difluoromethoxy)pyridin-3-yl)methanamine hydrochloride
Vzorec: C₇H₈F₂N₂O·ClH
Storage Temperature: Ambient
MDL Number: MFCD31621138
CAS číslo: 2231672-97-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR