Tisk…

4-(Difluoromethoxy)-5-fluoropyridin-2-amine

Dodavatel: Apollo Scientific
Varování

PC400139-250MG PC400139-100MG
APOSPC400139-250MGEA 22490 CZK
APOSPC400139-250MG APOSPC400139-100MG
4-(Difluoromethoxy)-5-fluoropyridin-2-amine
4-(Difluoromethoxy)-5-fluoropyridin-2-amine
Vzorec: C₆H₅F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD31621121
CAS číslo: 2231676-95-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR