Tisk…

5-Chloro-4-(difluoromethoxy)pyridin-2-amine

Dodavatel: Apollo Scientific
Varování

PC400138-250MG PC400138-100MG
APOSPC400138-250MGEA 22490 CZK
APOSPC400138-250MG APOSPC400138-100MG
5-Chloro-4-(difluoromethoxy)pyridin-2-amine
5-Chloro-4-(difluoromethoxy)pyridin-2-amine
Vzorec: C₆H₅ClF₂N₂O
MW: 194,567 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621119
CAS číslo: 2231672-98-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR