Tisk…

5-Chloro-4-(difluoromethoxy)pyridine-2-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Chloro-4-(difluoromethoxy)picolinicacid

PC400111-250MG PC400111-100MG
APOSPC400111-250MGEA 22490 CZK
APOSPC400111-250MG APOSPC400111-100MG
5-Chloro-4-(difluoromethoxy)pyridine-2-carboxylic acid
5-Chloro-4-(difluoromethoxy)pyridine-2-carboxylic acid
Vzorec: C₇H₄ClF₂NO₃
MW: 223.561 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751029
CAS číslo: 1805028-61-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR