(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanol
Dodavatel: Apollo Scientific
|
Varování
|
PC400108-100MG
PC400108-250MG
APOSPC400108-100MGEA
5200
CZK
APOSPC400108-100MG
APOSPC400108-250MG
(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanol
(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanol
Vzorec:
C₇H₆BrF₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD31621149 CAS číslo: 2231673-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...