Tisk…

(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanamine

Dodavatel: Apollo Scientific
Nebezpečí

PC400107-100MG PC400107-250MG
APOSPC400107-100MGEA 6190 CZK
APOSPC400107-100MG APOSPC400107-250MG
(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanamine
(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanamine
Vzorec: C₇H₇BrF₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD31621147
CAS číslo: 2231675-66-2
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR