Tisk…

1-(3-Bromo-5-chloropyridin-2-yl)-2,2,2-trifluoroethanone

Dodavatel: Apollo Scientific
Varování

PC400102-1G PC400102-250MG
APOSPC400102-1GEA 31080 CZK
APOSPC400102-1G APOSPC400102-250MG
1-(3-Bromo-5-chloropyridin-2-yl)-2,2,2-trifluoroethanone
1-(3-Bromo-5-chloropyridin-2-yl)-2,2,2-trifluoroethanone
Vzorec: C₇H₂BrClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD31621137
CAS číslo: 2231674-22-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR