Tisk…

1-(6-Chloro-5-methylpyridin-3-yl)-2,2,2-trifluoroethanone

Dodavatel: Apollo Scientific
Varování

PC400097-100MG PC400097-250MG
APOSPC400097-100MGEA 5200 CZK
APOSPC400097-100MG APOSPC400097-250MG
1-(6-Chloro-5-methylpyridin-3-yl)-2,2,2-trifluoroethanone
1-(6-Chloro-5-methylpyridin-3-yl)-2,2,2-trifluoroethanone
Vzorec: C₈H₅ClF₃NO
MW: 223.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29040610
CAS číslo: 1935574-78-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR