Tisk…

(2-Bromo-3,4-difluorophenyl)methanol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-3,4-difluorobenzyl alcohol

PC400044-250MG PC400044-1G
APOSPC400044-250MGEA 2070 CZK
APOSPC400044-250MG APOSPC400044-1G
(2-Bromo-3,4-difluorophenyl)methanol 95%
(2-Bromo-3,4-difluorophenyl)methanol
Vzorec: C₇H₅BrF₂O
MW: 223,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28657913
CAS číslo: 1578156-19-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR