Tisk…

tert-Butyl 2-bromo-3,4-difluorobenzylcarbamate

Dodavatel: Apollo Scientific
Nebezpečí

PC400038-100MG PC400038-250MG
APOSPC400038-100MGEA 5690 CZK
APOSPC400038-100MG APOSPC400038-250MG
tert-Butyl 2-bromo-3,4-difluorobenzylcarbamate
tert-Butyl 2-bromo-3,4-difluorobenzylcarbamate
Vzorec: C₁₂H₁₄BrF₂NO₂
MW: 322,146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695385
CAS číslo: 2231675-56-0
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR