Methyl 4-(4-methoxybenzylamino)-6-fluoropyridine-3-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 6-fluoro-4-((4-methoxybenzyl)amino)nicotinate
PC400033-1G
PC400033-250MG
APOSPC400033-1GEA
16200
CZK
APOSPC400033-1G
APOSPC400033-250MG
Methyl 4-(4-methoxybenzylamino)-6-fluoropyridine-3-carboxylate
Methyl 4-(4-methoxybenzylamino)-6-fluoropyridine-3-carboxylate
Vzorec:
C₁₅H₁₅FN₂O₃ MW: 290.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31536748 CAS číslo: 2187435-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...