Tisk…

5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-[(tert-Butyldimethylsilanyl)oxy]-2-fluorobenzaldehyde

PC400031-1G PC400031-5G
APOSPC400031-1GEA 2150 CZK
APOSPC400031-1G APOSPC400031-5G
5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde
5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde
Vzorec: C₁₃H₁₉FO₂Si
MW: 254.373 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17676576
CAS číslo: 113984-67-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR