Tisk…

2-Amino-4-bromo-3-fluorobenzonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-6-cyano-2-fluoroaniline

PC400006-1G PC400006-250MG PC400006-5G
APOSPC400006-1GEA 1210 CZK
APOSPC400006-1G APOSPC400006-250MG APOSPC400006-5G
2-Amino-4-bromo-3-fluorobenzonitrile
2-Amino-4-bromo-3-fluorobenzonitrile
Vzorec: C₇H₄BrFN₂
MW: 215,022 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26407501
CAS číslo: 1820620-31-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR