tert-Butyl 2-[4-(diphenylsulphonium)phenoxy] acetate, triflate salt
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC3992-1G
PC3992-5G
APOSPC3992-1GEA
1660
CZK
APOSPC3992-1G
APOSPC3992-5G
tert-Butyl 2-[4-(diphenylsulphonium)phenoxy] acetate, triflate salt
tert-Butyl 2-[4-(diphenylsulphonium)phenoxy] acetate, triflate salt
Photoinitiator/Photoacid Generator
Vzorec:
C₂₅H₂₅F₃O₆S₂ MW: 542,588 g/mol Bod tání: 106…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683567 CAS číslo: 180801-55-2 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...