Tisk…

[4-(tert-Butyl)phenyl](diphenyl)sulphonium trifluoromethanesulphonate

Dodavatel: Apollo Scientific
Varování

PC3895-1G PC3895-5G
APOSPC3895-1GEA 5200 CZK
APOSPC3895-1G APOSPC3895-5G
[4-(tert-Butyl)phenyl](diphenyl)sulphonium trifluoromethanesulphonate
[4-(tert-Butyl)phenyl](diphenyl)sulphonium trifluoromethanesulphonate

Photoinitiator/Photoacid Generator

Vzorec: C₂₂H₂₃S·CF₃O₃S
Bod tání: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD02683575
CAS číslo: 145612-66-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR