Quinolinium fluorochromate (QFC) on alumina
Dodavatel: Apollo Scientific
|
Varování
|
PC3876-25G
PC3876-5G
APOSPC3876-25GEA
14090
CZK
APOSPC3876-25G
APOSPC3876-5G
Quinolinium fluorochromate (QFC) on alumina
Quinolinium fluorochromate (QFC)
Selectively oxidises aliphatic primary and secondary alcohols and acyclic alcohols to their corresponding carbonyl compounds in good yield, ask for reference. Minimum activity 1mmol QFC per gram of alumina
Vzorec:
C₉H₇N·CrFHO₃ Storage Temperature: Ambient |
MDL Number:
MFCD02093330 CAS číslo: 143440-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...