2,6-Difluoro-4-(trifluoromethoxy)phenol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC37871-1G
PC37871-250MG
PC37871-5G
APOSPC37871-1GEA
16370
CZK
APOSPC37871-1G
APOSPC37871-250MG
APOSPC37871-5G
2,6-Difluoro-4-(trifluoromethoxy)phenol 95%
2,6-Difluoro-4-(trifluoromethoxy)phenol
Vzorec:
C₇H₃F₅O₂ MW: 214.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32219446 CAS číslo: 2383766-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...