2,6-Dichloro-4-fluorobenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC37870-1G
PC37870-250MG
PC37870-5G
APOSPC37870-1GEA
3970
CZK
APOSPC37870-1G
APOSPC37870-250MG
APOSPC37870-5G
2,6-Dichloro-4-fluorobenzenesulphonyl chloride
2,6-Dichloro-4-fluorobenzenesulphonyl chloride
Vzorec:
C₆H₂Cl₃FO₂S MW: 263,501 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16817657 CAS číslo: 88972-04-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...