Ethyl 6-nitro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
PC37862-1G
PC37862-250MG
APOSPC37862-1GEA
2690
CZK
APOSPC37862-1G
APOSPC37862-250MG
Ethyl 6-nitro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 6-nitro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Vzorec:
C₁₁H₈F₃N₃O₄ Storage Temperature: Ambient |
MDL Number:
MFCD32200424 CAS číslo: 2407339-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...