Tisk…

2-Bromo-3-fluorobenzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC3705-1G PC3705-250MG
APOSPC3705-1GEA 8770 CZK
APOSPC3705-1G APOSPC3705-250MG
2-Bromo-3-fluorobenzenesulphonyl chloride
2-Bromo-3-fluorobenzenesulphonyl chloride
Vzorec: C₆H₃BrClFO₂S
MW: 273,507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11101430
CAS číslo: 1065076-31-4
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR