HFPO polymer allyl ether
Dodavatel: Apollo Scientific
PC3632-1G
PC3632-25G
PC3632-5G
APOSPC3632-1GEA
2820
CZK
APOSPC3632-1G
APOSPC3632-25G
APOSPC3632-5G
HFPO polymer allyl ether
HFPO polymer allyl ether
(1500 - 2000 amu)
Storage Temperature:
Ambient |
MDL Number:
MFCD09753871 CAS číslo: 631897-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...