Tisk…

4-Methoxy-2,3,5,6-tetrafluorobenzyl alcohol 97%

Dodavatel: Apollo Scientific

PC3547-1G PC3547-250MG PC3547-5G
APOSPC3547-1GEA 12200 CZK
APOSPC3547-1G APOSPC3547-250MG APOSPC3547-5G
4-Methoxy-2,3,5,6-tetrafluorobenzyl alcohol 97%
4-Methoxy-2,3,5,6-tetrafluorobenzyl alcohol
Vzorec: C₈H₆F₄O₂
MW: 210,126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08741397
CAS číslo: 35175-79-2
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR