Tisk…

4-(Difluoromethoxy)phenol 97%

Dodavatel: Apollo Scientific
Varování

PC3536-1G PC3536-5G
APOSPC3536-1GEA 3770 CZK
APOSPC3536-1G APOSPC3536-5G
4-(Difluoromethoxy)phenol 97%
4-(Difluoromethoxy)phenol
Vzorec: C₇H₆F₂O₂
MW: 160,118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236236
CAS číslo: 87789-47-7
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR