Tisk…

2,3-Difluoro-4-(trifluoromethyl)benzyl alcohol 98%

Dodavatel: Apollo Scientific
Varování

PC3477-1G PC3477-250MG
APOSPC3477-1GEA 6220 CZK
APOSPC3477-1G APOSPC3477-250MG
2,3-Difluoro-4-(trifluoromethyl)benzyl alcohol 98%
2,3-Difluoro-4-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₅F₅O
Storage Temperature: Ambient
MDL Number: MFCD00236260
CAS číslo: 238403-50-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR