Tisk…

2-Fluorobenzal bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC3471-1G PC3471-5G
APOSPC3471-1GEA 5720 CZK
APOSPC3471-1G APOSPC3471-5G
2-Fluorobenzal bromide
2-Fluorobenzal bromide
Vzorec: C₇H₅Br₂F
MW: 267,921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045061
CAS číslo: 220141-76-4
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR