Tisk…

1-(2-Fluoroethyl)-4-hydroxypiperidine

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(2-Fluoroethyl)piperidin-4-ol

PC3464-1G PC3464-500MG
APOSPC3464-1GEA 23410 CZK
APOSPC3464-1G APOSPC3464-500MG
1-(2-Fluoroethyl)-4-hydroxypiperidine
1-(2-Fluoroethyl)-4-hydroxypiperidine
Vzorec: C₇H₁₄FNO
MW: 147,191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09751123
CAS číslo: 492446-45-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR