Tisk…

2-Bromo-6-fluoro-3-methoxybenzeneboronic acid

Dodavatel: Apollo Scientific

Synonyms: (2-Bromo-6-fluoro-3-methoxyphenyl)boronic acid

PC3439-100G PC3439-25G PC3439-5G
APOSPC3439-100GEA 16070 CZK
APOSPC3439-100G APOSPC3439-25G APOSPC3439-5G
2-Bromo-6-fluoro-3-methoxybenzeneboronic acid
2-Bromo-6-fluoro-3-methoxybenzeneboronic acid
Vzorec: C₇H₇BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD08276828
CAS číslo: 957062-89-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR