Pentafluorophenyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate 97%
Dodavatel: Apollo Scientific
PC3414-1G
PC3414-250MG
APOSPC3414-1GEA
5990
CZK
APOSPC3414-1G
APOSPC3414-250MG
Pentafluorophenyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate 97%
Pentafluorophenyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate
Vzorec:
C₁₇H₁₄F₅N₃O₂ MW: 387,304 g/mol Bod tání: 76,5…78.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD09879919 CAS číslo: 941716-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...