4-[(2-Fluoro-4-nitrophenyl)amino]piperidine, N1-BOC protected
Dodavatel: Apollo Scientific
Synonyms:
tert-Butyl 4-[(2-fluoro-4-nitrophenyl)amino]piperidine-1-carboxylate
PC3367-1G
PC3367-250MG
APOSPC3367-1GEA
5630
CZK
APOSPC3367-1G
APOSPC3367-250MG
4-[(2-Fluoro-4-nitrophenyl)amino]piperidine, N1-BOC protected
4-[(2-Fluoro-4-nitrophenyl)amino]piperidine, N1-BOC protected
Vzorec:
C₁₆H₂₂FN₃O₄ MW: 339,362 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09952122 CAS číslo: 952285-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...