Tisk…
4-[(4-Amino-2-fluorophenyl)(methyl)amino]piperidine, N1-BOC protected
Dodavatel: Apollo Scientific
Synonyms: tert-Butyl 4-[(4-amino-2-fluorophenyl)(methyl)amino]piperidine-1-carboxylate
PC3364-100MG PC3364-500MG
APOSPC3364-100MGEA 2900 CZK
APOSPC3364-100MG APOSPC3364-500MG
4-[(4-Amino-2-fluorophenyl)(methyl)amino]piperidine, N1-BOC protected
4-[(4-Amino-2-fluorophenyl)(methyl)amino]piperidine, N1-BOC protected
Vzorec: C₁₇H₂₆FN₃O₂
MW: 323.406 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09952121
CAS číslo: 952285-83-5